Anti-OLFM4 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP51203_T100
AVIVARP51203_T100EA 3850 NOK
AVIVARP51203_T100
Anti-OLFM4 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: OLFM4 (olfactomedin 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol OLFM4
Environmentally Preferable
Antigen-navn olfactomedin 4
Antigen synonymer GC1|OlfD|bA209J19.1|OLM4|GW112|UNQ362|hOLfD|hGC-1
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet
Vert Rabbit
Gen-ID 10562
Isotype
Western blot Yes
Epitop C-terminal
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR