Anti-C5orf55 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP52617_P050
AVIVARP52617_P050EA 4400 NOK
AVIVARP52617_P050
Anti-C5orf55 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: C5orf55 (chromosome 5 open reading frame 55)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol EXOC3-AS1
Antigen-navn EXOC3 antisense RNA 1
Environmentally Preferable
Antigen synonymer C5orf55
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human
Vert Rabbit
Gen-ID 116349
Isotype
Western blot Yes
Epitop N-terminal
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR