Anti-PRKAA1 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP53846_P050
AVIVARP53846_P050EA 4400 NOK
AVIVARP53846_P050
Anti-PRKAA1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PRKAA1 (protein kinase, AMP-activated, alpha 1 catalytic subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR