Anti-JAK3 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP54305_P050
AVIVARP54305_P050EA 4400 NOK
AVIVARP54305_P050
Anti-JAK3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: JAK3 (Janus kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol JAK3
Antigen-navn Janus kinase 3
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet
Vert Rabbit
Isotype
Western blot Yes
Epitop N-terminal
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR