Anti-CCDC110 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP55544_P050
AVIVARP55544_P050EA 4400 NOK
AVIVARP55544_P050
Anti-CCDC110 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CCDC110 (coiled-coil domain containing 110)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR