Anti-EML3 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP55579_P050
AVIVARP55579_P050EA 4400 NOK
AVIVARP55579_P050
Anti-EML3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: EML3 (echinoderm microtubule associated protein like 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol EML3
Antigen-navn echinoderm microtubule associated protein like 3
Environmentally Preferable
Antigen synonymer ELP95
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet
Vert Rabbit
Gen-ID 256364
Isotype
Western blot Yes
Epitop N-terminal
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR