Anti-FBXW8 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP55585_P050
AVIVARP55585_P050EA 4400 NOK
AVIVARP55585_P050
Anti-FBXW8 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: FBXW8 (F-box and WD repeat domain containing 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol FBXW8
Antigen-navn F-box and WD repeat domain containing 8
Environmentally Preferable
Antigen synonymer FBW8|FBXO29|FBXW6|FBW6|FBX29
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet
Vert Rabbit
Gen-ID 26259
Isotype
Western blot Yes
Epitop middle
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR