Anti-C4orf17 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP68042_P050
AVIVARP68042_P050EA 4400 NOK
AVIVARP68042_P050
Anti-C4orf17 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: C4orf17 (chromosome 4 open reading frame 17)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR