Anti-BRAF Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
OABF01143
AVIVOABF01143EA 7250 NOK
AVIVOABF01143
Anti-BRAF Rabbit Polyclonal Antibody
Antistoffer
Protein kinase involved in the transduction of mitogenic signals from the cell membrane to the nucleus. May play a role in the postsynaptic responses of hippocampal neuron. Phosphorylates MAP2K1, and thereby contributes to the MAP kinase signal transduction pathway.

Type:
Antigen: BRAF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, rat, human, dog, cow, pig, horse, chicken, rabbit
New Product
Bestill nå

SPESIFIKASJONER

Antigensymbol BRAF
Environmentally Preferable
Antigen-navn B-Raf proto-oncogene, serine/threonine kinase
Antigen synonymer B-Raf proto-oncogene serine/threonine-protein kinase (p94)|NS7|p94|proto-oncogene B-Raf|B-raf|B-Raf serine/threonine-protein|murine sarcoma viral (v-raf) oncogene homolog B1|RAFB1|94 kDa B-raf protein|v-raf murine sarcoma viral oncogene homolog B|serine/threonine-protein kinase B-raf|v-raf murine sarcoma viral oncogene homolog B1|BRAF1|B-RAF1
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Pig,Rabbit,Horse,Chicken,Human,Bovine,Dog,Rat,Mouse
Vert Rabbit
Gen-ID 673
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Immunogen KLH conjugated synthetic phosphopeptide derived from human B Raf around the phosphorylation site of Ser365
Format 0,01 M TBS (pH 7,4) with 1% BSA, 0,02% Proclin 300 and 50% glycerol
Skjema Liquid
Molekylvekt 84 kDa
Rensing Protein A purified
Konsentrasjon 1 µg/µl
Lagringstemp Store at –20 °C or 1 year. Avoid repeated freeze or thaw cycles.
UniProtKB P15056
Uttestet applikasjon ELISA, ICC, IF, IHC-Fr, IHC-P, WB

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR