Anti-PRDM1 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
P100596_P050
AVIVP100596_P050EA 4400 NOK
AVIVP100596_P050
Anti-PRDM1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PRDM1 (PR domain containing 1, with ZNF domain)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol PRDM1
Antigen-navn PR domain containing 1, with ZNF domain
Environmentally Preferable
Antigen synonymer PRDI-BF1|BLIMP1
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet
Vert Rabbit
Gen-ID 639
Isotype
Western blot Yes
Epitop middle
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR