Gel documentation systems, Axygen®
Leverandør: Corning
GD-1000 GDBL-1000
AXYGGD-1000EA 0 NOK
AXYGGD-1000 AXYGGDBL-1000
Gel documentation systems, Axygen®
Fotodokumentasjonssystemer
Axygen® gel documentation systems easily capture publication quality, 16 bit TIFF images. The systems are quick to set up and have an intuitive user interface for image capture, annotation, and contrast adjustment. Images are easily saved and opened in common gel analysis software for more detailed analysis.
  • Auto exposure - no manual manipulation of the camera required
  • Ready to use system - just connnect to computer and load the software
  • Long-life dual wavelength transilluminators
  • Exceptionally high resolution (5,4 MP) camera
  • UV cut-off switch to prevent