Axygen® Screw Caps for Transport Tubes
Leverandør: Corning
SC-B-510
AXYGSC-B-510EA 2525 NOK
AXYGSC-B-510
Axygen® Screw Caps for Transport Tubes
Lokk Lokk til reagensglass
These screw caps are used for 5/10 ml transport tubes.