Sedimentation cones, Imhoff type, Azlon®
Leverandør: SciLabware
SWX008
AZLOSWX008EA 3925 NOK
AZLOSWX008
Sedimentation cones, Imhoff type, Azlon®
Trakter Immhoff-trakter
Colourless cone, made up of transparent styrene-acrylonitrile S.A.N polymer.
  • With moulded graduations
  • Stopper at the bottom facilitates the removal of sample and cleaning
Sertifiseringer: Complies to DIN 12672