Holder for round bottom flasks
Leverandør: SciLabware
TWR038
AZLOTWR038EA 630 NOK
AZLOTWR038
Holder for round bottom flasks
Støtter Flaskeholdere
This stand is suitable for most round bottomed flasks.
  • Good chemical resistance
  • Stepped concentric rings to help stabilise flask