Autoclavable basket, Azlon®
Leverandør: SciLabware
TWR200
AZLOTWR200EA 585 NOK
AZLOTWR200
Autoclavable basket, Azlon®
Kurver Kurver, autklaverbare
PP, thanks to a robust and lightweight design, this autoclavable basket is suitable for draining applications as well as general storage.
  • Stackable when not used to save space
  • Good chemical resistance
  • Autoclavable to 121 °C
Colour: White.