MagBinder® Fit24 Nucleic Acid Purification System
Leverandør: OMEGA BIO-TEK
B1-001-24
B1-001-24EA 0 NOK
B1-001-24
MagBinder® Fit24 Nucleic Acid Purification System
Renseaparat for nuklein syrer
Elevate your laboratory experience with the MagBinder® Fit24 Nucleic acid purification system. Efficiency, affordability, precision, and reliability, all in one powerful instrument.
  • Ease-of-use – Automation-ready cartridges pre-filled with reagents
  • Convenience – No reagent preparation and buffer dispensing
  • High-quality purification – Up to 4 ml samples lysed offline eluted in as little as 50 μl
Simple and efficient – Experience efficient nucleic acid purification with the MagBinder® Fit24 Nucleic acid purification system. This affordable, state-of-the-More Product Information