trans-Aconitic acid 98

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: Trans-Propene-1,2,3-Tricarboxylic Acid, (E)-Prop-1-ene-1,2,3-tricarboxylic acid

B20087.14 B20087.22 B20087.PG
B20087.14EA 950 NOK
B20087.14 B20087.22 B20087.PG
trans-Aconitic acid 98
trans-Aconitic acid

Purity: 98% CAS No.: 4023-65-8 Molecular Formula: C6H6O6 Formula Weight: 174.11 Odour: Odourless Melting Point: ca 190° dec. MDL No.: MFCD00002721

Formel: C₆H₆O₆
MW: 174.11 g/mol
Smeltepunkt: ∼ 190 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002721
CAS-nummer: 4023-65-8
EINECS: 223-688-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR