4-(Trifluoromethylthio)benzaldehyde ≥90%

Leverandør: Thermo Fisher Scientific
Warning

B20313.03 B20313.06
B20313.03EA 780 NOK
B20313.03 B20313.06
4-(Trifluoromethylthio)benzaldehyde ≥90%
4-(Trifluoromethylthio)benzaldehyde

Purity: 90+% CAS No.: 4021-50-5 Molecular Formula: C8H5F3OS Formula Weight: 206.19 Flash Point: 96°(204°F) Refractive Index: 1.5110 Odour: Stench Melting Point: 29-31° Boiling Point: 219° Beilstein Reference: 2328411 MDL No.: MFCD00129191

Formel: C₈H₅F₃OS
Kokepunkt: 219 °C
Smeltepunkt: 29…31 °C
Tetthet: 1,310
Flash punkt: 96 °C (204 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00129191
CAS-nummer: 4021-50-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR