4-Methylthiobenzamide ≥97%

Leverandør: Thermo Fisher Scientific
Warning

B20320.03 B20320.06
B20320.03EA 395 NOK
B20320.03 B20320.06
4-Methylthiobenzamide ≥97%
4-Methylthiobenzamide

Purity: 97% CAS No.: 2362-62-1 Molecular Formula: C8H9NS Formula Weight: 151.23 Melting Point: 166-169° MDL No.: MFCD00173750

Formel: C₈H₉NS
Smeltepunkt: 166…169 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00173750
CAS-nummer: 2362-62-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR