1-Methylcycloheptanol ≥98%

Leverandør: Thermo Fisher Scientific
Warning

B20393.06
B20393.06EA 2225 NOK
B20393.06
1-Methylcycloheptanol ≥98%
1-Methylcycloheptanol

Purity: 98% CAS No.: 3761-94-2 Molecular Formula: C8H16O Formula Weight: 128.22 Refractive Index: 1.4720

Formel: C₈H₁₆O
Kokepunkt: 183,5 °C (1013 hPa)
Smeltepunkt: –4 °C
Tetthet: 0,918 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00021672
CAS-nummer: 3761-94-2
EINECS: 223-179-9

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR