2-Mercapto-5-methoxybenzimidazole ≥99%

Leverandør: Thermo Fisher Scientific
Warning

B20570.06 B20570.14 B20570.22
B20570.06EA 405 NOK
B20570.06 B20570.14 B20570.22
2-Mercapto-5-methoxybenzimidazole ≥99%
2-Mercapto-5-methoxybenzimidazole

Purity: 99% CAS No.: 37052-78-1 Molecular Formula: C8H8N2OS Formula Weight: 180.23 Melting Point: 255-259° Beilstein Reference: 3605123

Formel: C₈H₈N₂OS
Kokepunkt: 400 °C (1013 hPa)
Smeltepunkt: 255…259 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00134581
CAS-nummer: 37052-78-1
EINECS: 253-326-2

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR