Sodium deoxycholate monohydrate ≥98%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: Sodium 3alpha,12alpha-dihydroxy-5beta-cholanoate monohydrate, Deoxycholic acid sodium salt monohydrate, 3α,12α-Dihydroxycholan-24-oic acid sodium salt monohydrate, 3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt monohydrate, Sodium 3α,12α-dihydroxy-5β-cholanoate monohydrate, 3α,12α-Dihydroxy-5β-cholanic acid sodium salt monohydrate, 3α,12α-Dihydroxycholanic acid sodium salt monohydrate

B20759.14 B20759.22 B20759.36
B20759.14EA 915 NOK
B20759.14 B20759.22 B20759.36
Sodium deoxycholate monohydrate ≥98%
Sodium deoxycholate monohydrate

Purity: 98% CAS No.: 145224-92-6 Molecular Formula: C24H39NaO4.H2O Formula Weight: 432.58 (414.58anhy) Melting Point: 300° Beilstein Reference: 3643164 Merck Reference: 14,2899 Optical Rotation: +44° (c=2 in water) MDL No.: MFCD00150750

Formel: C₂₄H₃₉NaO₄·1H₂O
MW: 432.58 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00150750
CAS-nummer: 145224-92-6
EINECS: 206-132-7

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR