1,1'-Bis(diphenylphosphino)ferrocene ≥97%

Leverandør: Thermo Fisher Scientific
Warning

B21166.14
B21166.14EA 9525 NOK
B21166.14
1,1'-Bis(diphenylphosphino)ferrocene ≥97%
1,1'-Bis(diphenylphosphino)ferrocene

Purity: 97% CAS No.: 12150-46-8 Molecular Formula: C34H28FeP2 Formula Weight: 554.40 Storage Temperature: 2°C to 8°C Melting Point: ca 182° dec. MDL No.: MFCD00001422

Formel: C₃₄H₂₈FeP₂
Smeltepunkt: ∼ 182 °C (dec.)
MDL nummer: MFCD00001422
CAS-nummer: 12150-46-8
EINECS: 430-420-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR