Giemsa's stain for use in preparing solutions of leishman's, jenner's, giemsa's and other stains

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: Azure mixture sicc. Giemsa stain, Azur-Eosin-Methylene blue acc. Giemsa, Giemsa's azur-eosin-methylene blue

B21172.09 B21172.18
B21172.09EA 445 NOK
B21172.09 B21172.18
Giemsa's stain for use in preparing solutions of leishman's, jenner's, giemsa's and other stains
Giemsa's stain

Giemsa stain is a laboratory reagent and stain for microscopy used in cytogenetics and histopathologic detection of malaria and other parasites such as spirochete and protozoan blood parasites.


The stain can be incorporated as a differential staining between bacterial and human cells.


It can also be used to visualize chromosomes or to identify mast cells.

Smeltepunkt: ∼ 300 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00081642
CAS-nummer: 51811-82-6
EINECS: 257-438-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR