Resazurin sodium salt

Leverandør: Thermo Fisher Scientific
Warning

B21187.03 B21187.06
B21187.03EA 535 NOK
B21187.03 B21187.06
Resazurin sodium salt
Resazurin sodium salt

CAS No.: 62758-13-8 Molecular Formula: C12H6NNaO4 Formula Weight: 251.18 Physical Form: Powder Odour: Odourless Beilstein Reference: 3794971 Merck Reference: 14,8141 MDL No.: MFCD00005036


As indicator pH 3.8 (orange) to 6.5 (dark violet)

Formel: C₁₂H₆NNaO₄
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005036
CAS-nummer: 62758-13-8
EINECS: 263-718-5
Merck Indeks: 14,8141

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR