2,5-Dimethylstyrene ≥97% stabilised

Leverandør: Thermo Fisher Scientific
Warning


Stabilisering: Stabilised with 4-tert-butylpyrocatechol (TBC)

B21476.03 B21476.06 B21476.14
B21476.03EA 535 NOK
B21476.03 B21476.06 B21476.14
2,5-Dimethylstyrene ≥97% stabilised
2,5-Dimethylstyrene

Stabilized with 4-tert-butylcatechol


Purity: 97% CAS No.: 2039-89-6 Molecular Formula: C10H12 Formula Weight: 132.21 Storage Temperature: 2°C to 8°C Flash Point: 63°(145°F) Refractive Index: 1.5390 Melting Point: -35° Boiling Point: 83-84°/24mm Beilstein Reference: 2203656 MDL No.: MFCD00008614

Formel: C₁₀H₁₂
MW: 132.21 g/mol
Kokepunkt: 83…84 °C (24 mmHg)
Smeltepunkt: –35 °C
Tetthet: 0.904
Flash punkt: 63 °C (145 °F)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00008614
CAS-nummer: 2039-89-6
EINECS: 218-028-9

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR