Klorfenolrødt

Leverandør: Thermo Fisher Scientific
Warning

B21623.06 B21623.14
B21623.06EA 635 NOK
B21623.06 B21623.14
Klorfenolrødt
Klorfenolrødt

CAS No.: 4430-20-0 Molecular Formula: C19H12Cl2O5S Formula Weight: 432.28 Beilstein Reference: 354053 MDL No.: MFCD00005877

Formel: C₁₉H₁₂Cl₂O₅S
Smeltepunkt: ∼261 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005877
CAS-nummer: 4430-20-0
EINECS: 224-619-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR