Safranine O

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: CI 50240

B21674.09 B21674.18
B21674.09EA 420 NOK
B21674.09 B21674.18
Safranine O
Safranine O

CAS No.: 477-73-6 Molecular Formula: C20H19ClN4 Formula Weight: 350.85 Odour: Odourless Beilstein Reference: 3924099 MDL No.: MFCD00011759

Formel: C₂₀H₁₉ClN₄
MW: 350.85 g/mol
Tetthet: 1.447 g/cm³ (22 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011759
CAS-nummer: 477-73-6
EINECS: 207-518-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR