(Methylthio)acetaldehyde dimethyl acetal ≥97%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: (Methylthio)acetaldehydedimethylacetal

B22667.14
B22667.14EA 1825 NOK
B22667.14
(Methylthio)acetaldehyde dimethyl acetal ≥97%
(Methylthio)acetaldehyde dimethyl acetal

Purity: 97% CAS No.: 40015-15-4 Molecular Formula: CH3SCH2CH(OCH3)2 Formula Weight: 136.21 Flash Point: 53°(127°F) Refractive Index: 1.4528 Boiling Point: 170-171° Beilstein Reference: 773689 UN No.: 1993 MDL No.: MFCD00009848

Formel: C₅H₁₂O₂S
MW: 136.21 g/mol
Kokepunkt: 170…171 °C
Tetthet: 1.022
Flash punkt: 53 °C (127 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009848
CAS-nummer: 40015-15-4
ADR: 3 III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR