2-Chloro-p-toluidine ≥98%

Leverandør: Thermo Fisher Scientific
Danger

B22935.06 B22935.14
B22935.06EA 505 NOK
B22935.06 B22935.14
2-Chloro-p-toluidine ≥98%
2-Chloro-p-toluidine

Purity: 98% CAS No.: 615-65-6 Molecular Formula: C7H8ClN Formula Weight: 141.60 Flash Point: 99°(210°F) Refractive Index: 1.5750 Melting Point: 7° Boiling Point: 223-225° Beilstein Reference: 774514 UN No.: 3429 MDL No.: MFCD00007666

Formel: C₇H₈ClN
Kokepunkt: 223…225 °C
Smeltepunkt: 7 °C
Tetthet: 1,151
Flash punkt: 99 °C (210 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007666
CAS-nummer: 615-65-6
EINECS: 210-440-7
FN: 3429
ADR: 6.1,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR