Lys Grønn SF Gulaktig

Leverandør: Thermo Fisher Scientific
Warning

B23330.09 B23330.18
B23330.09EA 620 NOK
B23330.09 B23330.18
Lys Grønn SF Gulaktig
Lys Grønn SF Gulaktig

CAS No.: 5141-20-8 Molecular Formula: C37H34N2Na2O9S3 Formula Weight: 792.86 Melting Point: ca 288° dec. Beilstein Reference: 5717828 Merck Reference: 14,5485 MDL No.: MFCD00012121

Formel: C₃₇H₃₄N₂Na₂O₉S₃
Smeltepunkt: ∼ 288 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012121
CAS-nummer: 5141-20-8
EINECS: 225-906-5
Merck Indeks: 14,5485

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR