N,N'-Diphenyl-p-phenylenediamine 97%

Leverandør: Thermo Fisher Scientific
Warning

B23713.06 B23713.14 B23713.22
B23713.06EA 500 NOK
B23713.06 B23713.14 B23713.22
N,N'-Diphenyl-p-phenylenediamine 97%
N,N'-Diphenyl-p-phenylenediamine

Purity: 97% CAS No.: 74-31-7 Molecular Formula: C18H16N2 Formula Weight: 260.34 Melting Point: 146-153° Boiling Point: 220-225°/1mm Merck Reference: 14,3329 MDL No.: MFCD00003015

Formel: C₁₈H₁₆N₂
Kokepunkt: 220…225 °C (1 mmHg)
Smeltepunkt: 146…153 °C
Tetthet: 1,28
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003015
CAS-nummer: 74-31-7
EINECS: 200-806-4
Merck Indeks: 14,3329

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR