Zincon sodium salt

Leverandør: Thermo Fisher Scientific
Warning

B24256.03 B24256.06 B24256.14
B24256.03EA 535 NOK
B24256.03 B24256.06 B24256.14
Zincon sodium salt
Zincon sodium salt

CAS No.: 62625-22-3 Molecular Formula: C20H15N4NaO6S Formula Weight: 462.42 Melting Point: ca 203° dec. Beilstein Reference: 3872699 MDL No.: MFCD00064385

Formel: C₂₀H₁₅N₄NaO₆S
Smeltepunkt: ∼ 203 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064385
CAS-nummer: 62625-22-3
EINECS: 263-651-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR