N-Benzoylsuccinimide 97%

Leverandør: Thermo Fisher Scientific
Warning

B24680.06
B24680.06EA 1000 NOK
B24680.06
N-Benzoylsuccinimide 97%
N-Benzoylsuccinimide

Purity: 97% CAS No.: 6343-27-7 Molecular Formula: C9H9NO3 Formula Weight: 179.18 MDL No.: MFCD00185021

Formel: C₉H₉NO₃
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00185021
CAS-nummer: 6343-27-7

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR