Jern, high purity

Leverandør: Thermo Fisher Scientific

B25007.18 B25007.30
B25007.18EA 535 NOK
B25007.18 B25007.30
Jern, high purity
Jern
Formel: C₁₀H₁₀Fe
Kokepunkt: 249 °C (1013 hPa)
Smeltepunkt: 172…175 °C
Tetthet: 1,49 g/cm³ (20 °C)
MDL nummer: MFCD00001427
CAS-nummer: 102-54-5
EINECS: 203-039-3
FN: 1325
ADR: 4.1,II
Merck Indeks: 13,04070

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR