Leverandør: BANDELIN-ELECTRONIC
3763
BAND3763EA 5875 NOK
BAND3763
Temperaturprober
By connecting the temperature sensor either to the ultra sonic generator (series HD 4000) or to the ultrasonic converter (series HD 5000), temperature detection is activated and user-defined temperature monitoring is possible during the sonication process.
  • Sample temperatures in the range from –10 to +120 °C can be measured
  • High temperatures must not be allowed to enter the ultrasonic converter (max. 80 °C)
  • Long-term exposure to high temperatures must be avoided