Ultrasonic Bath with Heating, Sonorex Technology RM 182 UH
Leverandør: BANDELIN-ELECTRONIC
9202
BAND9202EA 0 NOK
BAND9202
Ultrasonic Bath with Heating, Sonorex Technology RM 182 UH
Ultralydbad
High-performance ultrasonic cleaning bath with heating for industry, service and maintenance. The welded oscillating tank made of stainless steel V4A (1 4404), 2 mm guarantees maximum durability even with intensive use.
  • Particularly durable due to welded oscillating tank
  • With sloping floor and round tub corners
  • Simple operation
  • With spray tube and overflow pocket
  • Modularly combinable to form a cleaning line
  • Fixed power setting
  • Operation: Rotary handle
  • Time setting: 1 to 15 minutes
  • Handles: On the sides
  • Shape of the tub corners: Rounded