Magnetic hotplate stirrers, RT2 basic
Leverandør: Thermo Fisher Scientific
88880003
BARN88880003EA 8050 NOK
BARN88880003
Magnetic hotplate stirrers, RT2 basic
Omrørere Magnetrørere med varmeplate
Basic hotplate is equipped with hot top warning and built in over temperature shut-off to provide accurate heating of samples and safety to the laboratory. Aluminum top provides excellent temperature uniformity while the thin ceramic coating provides excellent corrosion resistance and a white surface for optimal sample viewing.
  • Automatic Over-temperature safety shut off at 450 °C
  • Hot top warning indicator
  • Two threaded holes for support rods
Provided with CE and cCSAUS certificate.