Hotplate stirrers, digital, RT2 Advanced
Leverandør: Thermo Fisher Scientific
88880005
BARN88880005EA 10475 NOK
BARN88880005
Hotplate stirrers, digital, RT2 Advanced
Omrørere Magnetrørere med varmeplate
The RT2 is alow-profile digital display hotplatestirrer, equipped with PT100 sensor for precise temperature measurements for sensitive experimental procedures with built-in temperature shut-off to heat samples safely and accurately. The aluminium top provides excellent temperature uniformity while the thin ceramic coating provides excellent corrosion resistance and a white surface for optimal sample viewing. The unit features a keypad lock, hot top warning indicator and has two threaded holes to accommodate support rods.
  • Precise digital control with 0,1 ºC resolution
  • Microprocessor PID