Deionizer, barnstead Bantam™
Leverandør: Thermo Fisher Scientific
D0805
BARND0805EA 18450 NOK
BARND0805
Deionizer, barnstead Bantam™
Vannrensesystemer Ionebyttere
It is a simple, cost effective method for deionisation or pretreatment of lab water. Typically these units are ideal where water usage is up to 38 litres per day.