Cimarec+™ Hotplate
Leverandør: Thermo Fisher Scientific
HP88857108
BARNHP88857108EA 4675 NOK
BARNHP88857108
Cimarec+™ Hotplate
Termostater Varmeplater
The Thermo Scientific Cimarec+™ series digital hotplates are designed to provide exceptional safety, and temperature performance for routine protocols
All units offer digital displays of temperature. Temperature is adjustable in 1° increments. The HOT TOP warning system is a standard feature, and is designed to protect users from accidental burns. The HOT TOP feature provides a prominent display when heating surface is above 50 °C, even when unit is shut off (as long as the unit remains plugged in). The Cimarec+™ series also features automatic over-temperature protection, and a raisedMore Product Information
Bestill nå
SPESIFIKASJONER
Elektronisk 230 V, 50/60 Hz
Environmentally Preferable
Material toppanne Aluminum
Dimensjon toppanne 177,4×177,4 mm