Orbital shaker, benchtop, MaxQ™ HP
Leverandør: Thermo Fisher Scientific
SHKE416HP-1CE
BARNSHKE416HP-1CEEA 59700 NOK
BARNSHKE416HP-1CE
Orbital shaker, benchtop, MaxQ™ HP
Risteapparater Kretsende risteapparater
MaxQ™ HP benchtop orbital shaker suitable for environmental rooms or on lab benches. It features balancing technology and rugged mechanism make smooth high-capacity 24/7 shaking a reality, an industry-leading platform size, wide-range speed, low heat output motor and spill-resistant design. It features set point security which restricts unauthorised or inadvertent changes to key functions. Equipped with alarm to know the shaker performance.
  • Triple counter-balanced mechanism prevents vibration, allowing the unit to run smoothly with heavy, or out-of-balance