Incubating Shakers, MaxQ™ 8000
Leverandør: Thermo Fisher Scientific
SHKE8000-8CE SHKE8000-1CE SHKE8000-1CEPR
444-0897EA 0 NOK
444-0897 444-0896 BARNSHKE8000-1CEPR
Incubating Shakers, MaxQ™ 8000
Risteapparater Risteinkubatorer
With three times the capacity of a floor shaker in virtually the same footprint, these units are ideal for high volume research or scale-up work. They are built both for sensitive eukaryotic culture applications - with contamination-reducing HEPA filtration, coved corner design, single piece chamber, and tight temperature uniformity - and for demanding prokaryotic culture applications, with rugged construction, wide speed range, and no limits on 'hard shake' speed, even when stacked three-high.
  • Microprocessor control/monitoring system: Run and setpoints displayed simultaneously; tracks