DSP (Dithiobis(succinimidyl propionate))

Leverandør: G Bioscienes

Synonymer: DTSP

BC07
BC07EA 8700 NOK
BC07
DSP (Dithiobis(succinimidyl propionate))
DSP (Dithiobis(succinimidyl propionate))

Dithiobis(succinimidyl propionate) Homobifunctional Cross Linker.

Formel: C₁₄H₁₆N₂O₈S₂
MW: 404.41 g/mol
MDL nummer: MFCD00042045
CAS-nummer: 57757-57-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0236ade3279bb4cd4a60cb9dd756af9a0a277192&vid=0236ade3279bb4cd4a60cb9dd756af9a0a277192&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Spacer Arm (Å) 12
Reactive Toward Primary Amines
Membrane Permeable Yes
Water Soluble No
Cleavable/ Reversible Reducing Agents (Thiols)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR