Sulphosuccinimidyl acetate (Sulpho-NHS-acetate)

Leverandør: G Bioscienes

Synonymer: Sulpho-NHS-acetate, 1-(Acetyloxy)-2,5-dioxo-3-pyrrolidinesulphonic acid

BC91
BC91EA 6025 NOK
BC91
Sulphosuccinimidyl acetate (Sulpho-NHS-acetate)
Sulphosuccinimidyl acetate (Sulpho-NHS-acetate)

Sulpho NHS acetate is a small, water-soluble compound used for blocking primary amines by acylation. It covalent bonds with amine groups to prevent specific interactions and conjugations. It is water-soluble and reacts at pH 7 to 9.

Formel: C₄H₄NNaO₆S
MW: 259.17 g/mol
MDL nummer: MFCD01861959
CAS-nummer: 152305-87-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84eafb3d83cf3753f2f1f5929f70be73142165a5&vid=eaf30c81e4191e14bcffffeec9346569c2d693a6&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR