4-Hydroxyphenyl glyoxal (HPG)

Leverandør: G Bioscienes

Synonymer: HPG

BC94
BC94EA 8275 NOK
BC94
4-Hydroxyphenyl glyoxal (HPG)
4-Hydroxyphenyl glyoxal (HPG)

G-Biosciences' HPG (p-hydroxyphenylglyoxal) specifically reacts with arginine residues under mild conditions (pH 7 to 9, 25°C) to yield spectrophotometrically measurable signal for amino acid detection.


G-Biosciences' HPG is more water-soluble and more resistant to oxidation than other arginine modifiers. Modification can be monitored at 340 nm.

Formel: C₈H₆O₃
MW: 150.131 g/mol
MDL nummer: MFCD00054984
CAS-nummer: 24645-80-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ed3a4b861ac0738f0b1529c54ba0842e34099cc&vid=bfcd1a49bc114e3449607488ccd78a7a3d14c547&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR