2-Merkaptoetanol

Leverandør: G Bioscienes
Danger

Synonymer: 2-Hydroxyethyl mercaptan, β-Mercaptoethanol

BC98
BC98EA 1700 NOK
BC98
2-Merkaptoetanol
2-Merkaptoetanol
Formel: HSCH₂CH₂OH
MW: 78.14 g/mol
Kokepunkt: 157
Smeltepunkt: –100 °C
Tetthet: 1.12 g/cm³ (20 °C)
Flash punkt: 68
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004890
CAS-nummer: 60-24-2
EINECS: 200-464-6
FN: 2966
ADR: 6.1 II
Merck Indeks: 13,05894
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc427209b84d0343a5ae47c435ddfc53df4013d2&vid=7d70fd087d948d54e7a946d3b2310e2d820d2396&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR