Hybridoma growth and expression media HYGM6
Leverandør: BIOCONCEPT
9-00F55-I
BCON9-00F55-IEA 350 NOK
BCON9-00F55-I
Hybridoma growth and expression media HYGM6
Media, Cellekultur
Hybridoma growth media (HYGM) 6 is a serum-free and fully defined media that can be used for the cultivation of various different hybridomas and the production of monoclonal antibodies.
  • Serum-free
  • Ready to use medium
  • High yielding
  • contsins therapeutic grade recombinant insulin
  • Free of hydrolysates or peptones
  • Free of any undefined components
HYGM-6 medium contains recombinant therapeutic grade insulin. It is the only protein in that medium, no other proteins or undefined hydrolysates are present.
Sertifiseringer: ISO 9001; cGMP.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Sterilitet Sterile
Fraktforhold Ambient
Lagringstemp 2…4 °C