BD PrecisionGlide™ Needles
Leverandør: BD Medical
305196
BDAM305196EA 4050 NOK
BDAM305196
BD PrecisionGlide™ Needles
Sprøytenåler
BD PrecisionGlide™ Needle technology incorporates a thin wall, tri-bevel tip and a proprietary lubricant which is designed to provide ease of penetration.
  • Using a BD thin-wall needle may enable you to use a smaller gauge needle as the flow rate is similar to a BD regular-wall needle one gauge larger
  • BD has 20 years experience manufacturing thin-wall needles and meet ISO standards for cannula stiffness and resistance to breakage
  • 69% of patients said they would prefer to receive a skin injection with a smaller-diameter needle