Oralsprøyter, BD™
Leverandør: BD Medical
305207 305209 305210 305217 305219 305220
BDAM305207EA 4375 NOK
BDAM305207 BDAM305209 BDAM305210 BDAM305217 BDAM305219 BDAM305220
Oralsprøyter, BD™
Sprøyter Sprøyter, medisinsk
BD® oral/enteral syringe with BD UniVia™ connection is intended for use by healthcare professionals to measure and administer oral medication and enteral nutrition.
  • Non-luer compatible BD UniVia™ connection designed to reduce the risk of tubing set misconnections
  • Amber coloured syringes protect light-sensitive drugs
  • Oral syringes are used for the transport and administration of non-injectable liquid medication
  • Can be used in paediatric departments
  • Natural rubber latex, BPS, or DEHP are not part of the material formulation
  • Non-sterile
The plunger rodMore Product Information